Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Methyl 4-methoxy-3-nitrobenzoate, 97%

Click to view available options
Quantity:
1g
5g
25g
40757-20-8
C9H9NO5
211.173
MFCD00024213
ZUZYMTBOKNSYEB-UHFFFAOYSA-N
benzoic acid, 4-methoxy-3-nitro-, methyl ester,3-nitro-4-methoxybenzoic acid methyl ester,methyl 3-nitro-p-anisate,acmc-1ap03,methyl 3-nitro-4-methoxybenzoate,methyl-4-methoxy-3-nitrobenzoate,3-nitro-p-anisic acid methyl ester,4-methoxy-3-nitrobenzoic acid methyl ester,4-methoxy-3-nitro-benzoic acid methyl ester,benzoic acid, 4-methoxy-5-nitro-, methyl ester
602104
methyl 4-methoxy-3-nitrobenzoate
COC1=C(C=C(C=C1)C(=O)OC)[N+](=O)[O-]
40757-20-8
C9H9NO5
211.173
MFCD00024213
ZUZYMTBOKNSYEB-UHFFFAOYSA-N
benzoic acid, 4-methoxy-3-nitro-, methyl ester,3-nitro-4-methoxybenzoic acid methyl ester,methyl 3-nitro-p-anisate,acmc-1ap03,methyl 3-nitro-4-methoxybenzoate,methyl-4-methoxy-3-nitrobenzoate,3-nitro-p-anisic acid methyl ester,4-methoxy-3-nitrobenzoic acid methyl ester,4-methoxy-3-nitro-benzoic acid methyl ester,benzoic acid, 4-methoxy-5-nitro-, methyl ester
602104
methyl 4-methoxy-3-nitrobenzoate
COC1=C(C=C(C=C1)C(=O)OC)[N+](=O)[O-]
Melting Point 109°C to 112°C
Quantity 5g
Formula Weight 211.18
Percent Purity 97%
Chemical Name or Material Methyl 4-methoxy-3-nitrobenzoate