Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 4-methoxyphenylacetate, 97+%, Thermo Scientific Chemicals

Catalog Number AAA1794014
Click to view available options
Quantity:
25 g
100 g
500 g
23786-14-3
C10H12O3
180.20
MFCD00008454
ZQYLDVNTWDEAJI-UHFFFAOYSA-N
methyl 4-methoxyphenylacetate, methyl 2-4-methoxyphenyl acetate, methyl 4-methoxyphenyl acetate, 4-methoxyphenylacetic acid methyl ester, 4-methoxy-phenyl-acetic acid methyl ester, methyl 4-methoxybenzeneacetate, methyl p-methoxyphenyl acetate, benzeneacetic acid, 4-methoxy-, methyl ester, acetic acid, p-methoxyphenyl-, methyl ester, methyl 2-p-methoxyphenyl acetate
90266
methyl 2-(4-methoxyphenyl)acetate
COC(=O)CC1=CC=C(OC)C=C1
23786-14-3
C10H12O3
180.20
MFCD00008454
ZQYLDVNTWDEAJI-UHFFFAOYSA-N
methyl 4-methoxyphenylacetate, methyl 2-4-methoxyphenyl acetate, methyl 4-methoxyphenyl acetate, 4-methoxyphenylacetic acid methyl ester, 4-methoxy-phenyl-acetic acid methyl ester, methyl 4-methoxybenzeneacetate, methyl p-methoxyphenyl acetate, benzeneacetic acid, 4-methoxy-, methyl ester, acetic acid, p-methoxyphenyl-, methyl ester, methyl 2-p-methoxyphenyl acetate
90266
methyl 2-(4-methoxyphenyl)acetate
COC(=O)CC1=CC=C(OC)C=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 1.135
Boiling Point 157°C to 159°C (19 mmHg)
Flash Point >100°C (212°F)
Refractive Index 1.516
Quantity 25 g
Beilstein 2209665
Formula Weight 180.23
Percent Purity ≥97%
Chemical Name or Material Methyl 4-methoxyphenylacetate
Hazard Category H226
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: III
EINECSNumber 245-886-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit