Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Methyl 4-methyl-3-nitrobenzoate, 99%

Manufacturer:  Alfa Aesar™ B2529906

Quantity 5g

 View more versions of this product

Catalog No. AAB2529906


  Check Availability
Add to cart

Specifications

Description

Applications
Methyl 4-methyl-3-nitrobenzoate is used as pharmaceutical intermediate.

Solubility
Soluble in methanol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Description & Specifications

Chemical Identifiers

7356-11-8
195.18
4-Methyl-3-nitrobenzoic acid methyl ester; Methyl 3-nitro-p-toluate
C9H9NO4
MFCD00130054
Methyl 4-methyl-3-nitrobenzoate

Specifications

7356-11-8
195.18
4-Methyl-3-nitrobenzoic acid methyl ester; Methyl 3-nitro-p-toluate
5g
99%
C9H9NO4
MFCD00130054
49° to 53°C
Methyl 4-methyl-3-nitrobenzoate

Safety and Handling

SDS & Safety Documents

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit