Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Methyl 4-methyl-3-nitrobenzoate, 99%

Click to view available options
Quantity:
5g
25g
100g
7356-11-8
C9H9NO4
195.17
MFCD00130054
YFPBHPCMYFCRKS-UHFFFAOYSA-N
methyl 3-nitro-4-methylbenzoate,4-methyl-3-nitrobenzoic acid methyl ester,benzoic acid, 4-methyl-3-nitro-, methyl ester,methyl 3-nitro-p-toluate,4-methyl-3-nitro-benzoic acid methyl ester,pubchem10934,acmc-1bfci,ksc497g4t,methyl4-methyl-3-nitrobenzoate,4-methoxycarbonyl-2-nitrotoluene
81816
methyl 4-methyl-3-nitrobenzoate
CC1=C(C=C(C=C1)C(=O)OC)[N+](=O)[O-]
7356-11-8
C9H9NO4
195.17
MFCD00130054
YFPBHPCMYFCRKS-UHFFFAOYSA-N
methyl 3-nitro-4-methylbenzoate,4-methyl-3-nitrobenzoic acid methyl ester,benzoic acid, 4-methyl-3-nitro-, methyl ester,methyl 3-nitro-p-toluate,4-methyl-3-nitro-benzoic acid methyl ester,pubchem10934,acmc-1bfci,ksc497g4t,methyl4-methyl-3-nitrobenzoate,4-methoxycarbonyl-2-nitrotoluene
81816
methyl 4-methyl-3-nitrobenzoate
CC1=C(C=C(C=C1)C(=O)OC)[N+](=O)[O-]
Applications
Methyl 4-methyl-3-nitrobenzoate is used as pharmaceutical intermediate.

Solubility
Soluble in methanol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Melting Point 49°C to 53°C
Quantity 25g
Formula Weight 195.18
Percent Purity 99%
Chemical Name or Material Methyl 4-methyl-3-nitrobenzoate