Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Methyl 4-methyl-3-nitrobenzoate, 99%

Click to view available options
Quantity:
5g
25g
100g
7376-03-6
C23H14O
306.36
MFCD00130054
FGCOCPAQFSREBF-UHFFFAOYSA-N
4-Methyl-3-nitrobenzoic acid methyl ester; Methyl 3-nitro-p-toluate
13012745
phenyl(pyren-1-yl)methanone
C1=CC=C(C=C1)C(=O)C2=C3C=CC4=CC=CC5=C4C3=C(C=C5)C=C2
7376-03-6
C23H14O
306.36
MFCD00130054
FGCOCPAQFSREBF-UHFFFAOYSA-N
4-Methyl-3-nitrobenzoic acid methyl ester; Methyl 3-nitro-p-toluate
13012745
phenyl(pyren-1-yl)methanone
C1=CC=C(C=C1)C(=O)C2=C3C=CC4=CC=CC5=C4C3=C(C=C5)C=C2
Applications
Methyl 4-methyl-3-nitrobenzoate is used as pharmaceutical intermediate.

Solubility
Soluble in methanol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Melting Point 49°C to 53°C
Quantity 100g
Formula Weight 195.18
Percent Purity 99%
Chemical Name or Material Methyl 4-methyl-3-nitrobenzoate