Alfa Aesar™ Methyl 4'-methylbiphenyl-4-carboxylate, 95%

Manufacturer: Alfa Aesar™ H5191103

Quantity 1g

 View more versions of this product

Catalog No. AAH5191103

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 49742-56-5
    Molecular Formula C15H14O2
    Formula Weight 226.27
    MDL Number MFCD05979484
    Synonym 4'-Methylbiphenyl-4-carboxylic acid methyl ester
    Chemical Name or Material Methyl 4'-methylbiphenyl-4-carboxylate

    Specifications

    Formula Weight 226.27
    TSCA TSCA
    Molecular Formula C15H14O2
    MDL Number MFCD05979484
    CAS 49742-56-5
    Synonym 4'-Methylbiphenyl-4-carboxylic acid methyl ester
    Quantity 1g
    Chemical Name or Material Methyl 4'-methylbiphenyl-4-carboxylate
    Assay 95%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit