Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Methyl 4-methylsalicylate, ≥98%

Click to view available options
Quantity:
5g
25g
4670-56-8
C9H10O3
166.176
MFCD00020130
UITFCFWKYAOJEJ-UHFFFAOYSA-N
methyl 4-methylsalicylate,methyl-4-methylsalicylate,benzoic acid, 2-hydroxy-4-methyl-, methyl ester,4-methylsalicylic acid methyl ester,methyl-4-methyl salicylate,methyl m-cresotate,2-hydroxy-4-methylbenzoic acid methyl ester,methyl 2-hydroxy-4-methyl-benzoate,2-hydroxy-4-methyl-benzoic acid methyl ester,methyl methylsalicylate
78400
methyl 2-hydroxy-4-methylbenzoate
CC1=CC(=C(C=C1)C(=O)OC)O
4670-56-8
C9H10O3
166.176
MFCD00020130
UITFCFWKYAOJEJ-UHFFFAOYSA-N
methyl 4-methylsalicylate,methyl-4-methylsalicylate,benzoic acid, 2-hydroxy-4-methyl-, methyl ester,4-methylsalicylic acid methyl ester,methyl-4-methyl salicylate,methyl m-cresotate,2-hydroxy-4-methylbenzoic acid methyl ester,methyl 2-hydroxy-4-methyl-benzoate,2-hydroxy-4-methyl-benzoic acid methyl ester,methyl methylsalicylate
78400
methyl 2-hydroxy-4-methylbenzoate
CC1=CC(=C(C=C1)C(=O)OC)O
Melting Point 27°C to 28°C
Boiling Point 242°C (literature)
Density 1.147g/mL
Color Brown to Yellow
Refractive Index 1.5370
Quantity 5g
Beilstein 2690247
Formula Weight 166.18
Physical Form Low Melting Solid
Percent Purity ≥98%
Chemical Name or Material Methyl 4-methylsalicylate
Show More Show Less

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : 2° to 8°C