Alfa Aesar™ Methyl 4-tert-butylphenylacetate, 97%

Manufacturer: Alfa Aesar™ A1242122

Quantity 100g

 View more versions of this product

Catalog No. AAA1242122

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 3549-23-3
    Molecular Formula C13H18O2
    Formula Weight 206.29
    MDL Number MFCD00051913
    Beilstein 2502880
    Synonym 4-tert-Butylphenylacetic acid methyl ester
    Chemical Name or Material Methyl 4-tert-butylphenylacetate

    Specifications

    Formula Weight 206.29
    Molecular Formula C13H18O2
    Flash Point >100°C (212°F)
    MDL Number MFCD00051913
    CAS 3549-23-3
    Beilstein 2502880
    Boiling Point 149° to 151°C (30mmHg)
    TSCA TSCA
    Synonym 4-tert-Butylphenylacetic acid methyl ester
    Quantity 100g
    Chemical Name or Material Methyl 4-tert-butylphenylacetate
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit