Learn More
Methyl 4-vinylbenzoate, 96%, Thermo Scientific Chemicals
Description
Methyl 4-vinylbenzoate is used as a fluid component for enhancing ink stability in printing and also in preparing vinyl substituted beta-diketones. Further, it is used as an organic intermediate for pharmaceuticals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Methyl 4-vinylbenzoate is used as a fluid component for enhancing ink stability in printing and also in preparing vinyl substituted beta-diketones. Further, it is used as an organic intermediate for pharmaceuticals.
Notes
Incompatible with oxidizing agents.
Specifications
Melting Point | 32°C to 37°C |
Quantity | 1 g |
Formula Weight | 162.19 |
Percent Purity | 97% |
Chemical Name or Material | Methyl 4-vinylbenzoate |
Safety and Handling
Hazard Category | H315-H319-H335 |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.