Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Methyl 5-acetylsalicylate, 98%

Manufacturer:  Alfa Aesar™ A1021522

Quantity 100g

 View more versions of this product

Catalog No. AAA1021522


  Check Availability
Add to cart

Specifications

Description

Applications
Methyl 5-acetylsalicylate is used in food flavorings and preservatives. It is useful in the relief of headache and muscle and joint aches It is also effective in reducing fever, inflammation, and swelling and thus has been used for treatment of rheumatoid arthritis, rheumatic fever, and mild infection.

Solubility
Soluble in methanol.

Notes
Incompatible with oxidizing agents. Store in a cool, dry condition in well sealed containers.
Description & Specifications

Chemical Identifiers

16475-90-4
194.19
2643824
Methyl 5-acetylsalicylate
C10H10O4
MFCD00040924
5-Acetylsalicylic acid methyl ester; Methyl 2-hydroxy-5-acetylbenzoate

Specifications

16475-90-4
194.19
2643824
58° to 62°C
Methyl 5-acetylsalicylate
C10H10O4
MFCD00040924
5-Acetylsalicylic acid methyl ester; Methyl 2-hydroxy-5-acetylbenzoate
100g
98%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit