Alfa Aesar™ Methyl 5-acetylsalicylate, 98%

Manufacturer: Alfa Aesar™ A1021514

Quantity 25g

 View more versions of this product

Catalog No. AAA1021514

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 16475-90-4
    Molecular Formula C10H10O4
    Formula Weight 194.19
    MDL Number MFCD00040924
    Beilstein 2643824
    Synonym 5-Acetylsalicylic acid methyl ester; Methyl 2-hydroxy-5-acetylbenzoate
    Chemical Name or Material Methyl 5-acetylsalicylate

    Specifications

    Formula Weight 194.19
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 16475-90-4
    Beilstein 2643824
    Health Hazard 1 Warning
    Melting Point 58° to 62°C
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Molecular Formula C10H10O4
    MDL Number MFCD00040924
    Synonym 5-Acetylsalicylic acid methyl ester; Methyl 2-hydroxy-5-acetylbenzoate
    Quantity 25g
    Chemical Name or Material Methyl 5-acetylsalicylate
    Assay 98%

    Description

    Applications
    Methyl 5-acetylsalicylate is used in food flavorings and preservatives. It is useful in the relief of headache and muscle and joint aches It is also effective in reducing fever, inflammation, and swelling and thus has been used for treatment of rheumatoid arthritis, rheumatic fever, and mild infection.

    Solubility
    Soluble in methanol.

    Notes
    Incompatible with oxidizing agents. Store in a cool, dry condition in well sealed containers.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit