Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 5-acetylsalicylate, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
16475-90-4
C10H10O4
194.186
MFCD00040924
XLSMGNNWSRNTIQ-UHFFFAOYSA-N
methyl 5-acetylsalicylate,5-acetylsalicylic acid methyl ester,5-acetyl-2-hydroxybenzoic acid methyl ester,methyl 2-hydroxy-5-acetylbenzoate,benzoic acid, 5-acetyl-2-hydroxy-, methyl ester,5-acetyl methyl salicylate,methyl 5-acethylsalicylate,pubchem22504,methyl5-acetylsalicylate,methyl-5-acetylsalicylate
85444
methyl 5-acetyl-2-hydroxybenzoate
CC(=O)C1=CC(=C(C=C1)O)C(=O)OC
16475-90-4
C10H10O4
194.186
MFCD00040924
XLSMGNNWSRNTIQ-UHFFFAOYSA-N
methyl 5-acetylsalicylate,5-acetylsalicylic acid methyl ester,5-acetyl-2-hydroxybenzoic acid methyl ester,methyl 2-hydroxy-5-acetylbenzoate,benzoic acid, 5-acetyl-2-hydroxy-, methyl ester,5-acetyl methyl salicylate,methyl 5-acethylsalicylate,pubchem22504,methyl5-acetylsalicylate,methyl-5-acetylsalicylate
85444
methyl 5-acetyl-2-hydroxybenzoate
CC(=O)C1=CC(=C(C=C1)O)C(=O)OC
Quantity 25g
Packaging Glass bottle
Infrared Spectrum Authentic
Color Undesignated
Melting Point 58°C to 62°C
Assay Percent Range 98%
Linear Formula CH3C(O)C6H3(OH)COOCH3
Formula Weight 194.19
Physical Form Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material Methyl 5-acetylsalicylate, 98%
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 240-532-2