Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Methyl 5-bromo-2-chloronicotinate, 98%

Click to view available options
Quantity:
5g
25g
100g
78686-79-0
C7H5BrClNO2
250.476
MFCD03844848
MOMQDEDQGJAKII-UHFFFAOYSA-N
methyl 5-bromo-2-chloronicotinate,methyl5-bromo-2-chloronicotinate,5-bromo-2-chloro-nicotinic acid methyl ester,methyl 5-bromo-2-chloro-3-pyridinecarboxylate,5-bromo-2-chloronicotinic acid methyl ester,3-pyridinecarboxylic acid, 5-bromo-2-chloro-, methyl ester,methyl 5-bromo-2-chloro-pyridine-3-carboxylate,5-bromo-2-chloro-3-methoxycarbonyl pyridine,pubchem12966,acmc-209pf0
2763343
methyl 5-bromo-2-chloropyridine-3-carboxylate
COC(=O)C1=CC(=CN=C1Cl)Br
78686-79-0
C7H5BrClNO2
250.476
MFCD03844848
MOMQDEDQGJAKII-UHFFFAOYSA-N
methyl 5-bromo-2-chloronicotinate,methyl5-bromo-2-chloronicotinate,5-bromo-2-chloro-nicotinic acid methyl ester,methyl 5-bromo-2-chloro-3-pyridinecarboxylate,5-bromo-2-chloronicotinic acid methyl ester,3-pyridinecarboxylic acid, 5-bromo-2-chloro-, methyl ester,methyl 5-bromo-2-chloro-pyridine-3-carboxylate,5-bromo-2-chloro-3-methoxycarbonyl pyridine,pubchem12966,acmc-209pf0
2763343
methyl 5-bromo-2-chloropyridine-3-carboxylate
COC(=O)C1=CC(=CN=C1Cl)Br
Quantity 5g
Melting Point 48°C to 53°C
Sensitivity Light, Air & Moisture Sensitive
Beilstein 4394169
Formula Weight 250.48
Physical Form Solid
Percent Purity 98%
Chemical Name or Material Methyl 5-bromo-2-chloronicotinate