Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Methyl 5-bromo-6-chloronicotinate, 98%

Click to view available options
Quantity:
1g
5g
25g
78686-77-8
C7H5BrClNO2
250.476
MFCD03844855
WINGWVOUOFMOJQ-UHFFFAOYSA-N
methyl 5-bromo-6-chloronicotinate,5-bromo-6-chloro-nicotinic acid methyl ester,methyl-5-bromo-6-chloronicotinate,3-pyridinecarboxylic acid, 5-bromo-6-chloro-, methyl ester,methyl 3-bromo-2-chloropyridine-5-carboxylate,zlchem 1175,3-bromo-4-chloro-benzoicacidmethylester,pubchem12980,acmc-1bdei,methyl5-bromo-6-chloronicotinate
2764313
methyl 5-bromo-6-chloropyridine-3-carboxylate
COC(=O)C1=CC(=C(N=C1)Cl)Br
78686-77-8
C7H5BrClNO2
250.476
MFCD03844855
WINGWVOUOFMOJQ-UHFFFAOYSA-N
methyl 5-bromo-6-chloronicotinate,5-bromo-6-chloro-nicotinic acid methyl ester,methyl-5-bromo-6-chloronicotinate,3-pyridinecarboxylic acid, 5-bromo-6-chloro-, methyl ester,methyl 3-bromo-2-chloropyridine-5-carboxylate,zlchem 1175,3-bromo-4-chloro-benzoicacidmethylester,pubchem12980,acmc-1bdei,methyl5-bromo-6-chloronicotinate
2764313
methyl 5-bromo-6-chloropyridine-3-carboxylate
COC(=O)C1=CC(=C(N=C1)Cl)Br
Melting Point 76°C to 79°C
Quantity 1g
Beilstein 4403950
Formula Weight 250.48
Physical Form Solid
Percent Purity 98%
Chemical Name or Material Methyl 5-bromo-6-chloronicotinate

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : 2° to 8°C