Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Methyl 5-bromoorotate, 97%

Click to view available options
Quantity:
250mg
1g
5g
30825-88-8
C6H5BrN2O4
249.02
MFCD07023034
UJCPEPQYSPRWGU-UHFFFAOYSA-N
methyl 5-bromo-2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylate,methyl 5-bromo-2,6-dioxo-1,3-dihydropyrimidine-4-carboxylate,methyl 5-bromo-2,6-dihydroxypyrimidine-4-carboxylate,4-pyrimidinecarboxylic acid, 5-bromo-1,2,3,6-tetrahydro-2,6-dioxo-, methyl ester
6324367
methyl 5-bromo-2,4-dioxo-1H-pyrimidine-6-carboxylate
COC(=O)C1=C(C(=O)NC(=O)N1)Br
30825-88-8
C6H5BrN2O4
249.02
MFCD07023034
UJCPEPQYSPRWGU-UHFFFAOYSA-N
methyl 5-bromo-2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylate,methyl 5-bromo-2,6-dioxo-1,3-dihydropyrimidine-4-carboxylate,methyl 5-bromo-2,6-dihydroxypyrimidine-4-carboxylate,4-pyrimidinecarboxylic acid, 5-bromo-1,2,3,6-tetrahydro-2,6-dioxo-, methyl ester
6324367
methyl 5-bromo-2,4-dioxo-1H-pyrimidine-6-carboxylate
COC(=O)C1=C(C(=O)NC(=O)N1)Br
Quantity 250mg
Melting Point 252°C to 254°C
Beilstein 915036
Formula Weight 249.02
Physical Form Solid
Percent Purity 97%
Chemical Name or Material Methyl 5-bromoorotate