Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Methyl 5-bromopyridine-2-carboxylate, 98+%

Click to view available options
Quantity:
1g
5g
29682-15-3
C7H6BrNO2
216.034
MFCD04112493
JEURNBCYNWNADN-UHFFFAOYSA-N
methyl 5-bromopicolinate,5-bromopyridine-2-carboxylic acid methyl ester,methyl 5-bromo-2-pyridinecarboxylate,2-pyridinecarboxylic acid, 5-bromo-, methyl ester,5-bromo-2-pyridinecarboxylic acid methyl ester,5-bromo-pyridine-2-carboxylic acid methyl ester,methyl5-bromopicolinate,pubchem10589,5-bromopyridine-2-carboxylicacidmethylester,acmc-1cjr7
7016458
methyl 5-bromopyridine-2-carboxylate
COC(=O)C1=NC=C(C=C1)Br
29682-15-3
C7H6BrNO2
216.034
MFCD04112493
JEURNBCYNWNADN-UHFFFAOYSA-N
methyl 5-bromopicolinate,5-bromopyridine-2-carboxylic acid methyl ester,methyl 5-bromo-2-pyridinecarboxylate,2-pyridinecarboxylic acid, 5-bromo-, methyl ester,5-bromo-2-pyridinecarboxylic acid methyl ester,5-bromo-pyridine-2-carboxylic acid methyl ester,methyl5-bromopicolinate,pubchem10589,5-bromopyridine-2-carboxylicacidmethylester,acmc-1cjr7
7016458
methyl 5-bromopyridine-2-carboxylate
COC(=O)C1=NC=C(C=C1)Br
Applications
Methyl 5-bromopyridine-2-carboxylate is used as an organic chemical synthesis intermediate.

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Quantity 1g
Melting Point 99°C to 103°C
Formula Weight 216.03
Percent Purity ≥98%
Chemical Name or Material Methyl 5-bromopyridine-2-carboxylate

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning