Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Methyl 5-bromosalicylate, ≥98%

Click to view available options
Quantity:
5g
25g
4068-76-2
C8H7BrO3
231.045
MFCD00016460
FJYDBKPPGRZSOZ-UHFFFAOYSA-N
methyl 5-bromosalicylate,benzoic acid, 5-bromo-2-hydroxy-, methyl ester,5-bromosalicylic acid methyl ester,methyl 5-bromo-2-hydroxybenzenecarboxylate,methyl 2-hydroxy-5-bromobenzoate,5-bromo-2-hydroxy-benzoic acid methyl ester,methyl5-bromosalicylate,pubchem19740,acmc-1afpq,methyl-5-bromosalicylate
77683
methyl 5-bromo-2-hydroxybenzoate
COC(=O)C1=C(C=CC(=C1)Br)O
4068-76-2
C8H7BrO3
231.045
MFCD00016460
FJYDBKPPGRZSOZ-UHFFFAOYSA-N
methyl 5-bromosalicylate,benzoic acid, 5-bromo-2-hydroxy-, methyl ester,5-bromosalicylic acid methyl ester,methyl 5-bromo-2-hydroxybenzenecarboxylate,methyl 2-hydroxy-5-bromobenzoate,5-bromo-2-hydroxy-benzoic acid methyl ester,methyl5-bromosalicylate,pubchem19740,acmc-1afpq,methyl-5-bromosalicylate
77683
methyl 5-bromo-2-hydroxybenzoate
COC(=O)C1=C(C=CC(=C1)Br)O
Melting Point 61°C to 65°C (literature)
Quantity 5g
Color Brown to White
Beilstein 3047854
Formula Weight 231.05
Physical Form Solid
Percent Purity ≥98%
Chemical Name or Material Methyl 5-bromosalicylate

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning