Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Methyl 5-bromovalerate, 97%

Click to view available options
Quantity:
5g
25g
100g
5454-83-1
C6H11BrO2
195.056
MFCD00000265
RAVVJKCSZXAIQP-UHFFFAOYSA-N
methyl 5-bromovalerate,5-bromovaleric acid methyl ester,pentanoic acid, 5-bromo-, methyl ester,methyl .omega.-bromopentanoate,5-bromopentanoic acid methyl ester,5-bromo-pentanoic acid methyl ester,methyl omega-bromopentanoate,methyl5-bromovalerate,valeric acid, 5-bromo-, methyl ester,methyl-5-bromovalerate
79557
methyl 5-bromopentanoate
COC(=O)CCCCBr
5454-83-1
C6H11BrO2
195.056
MFCD00000265
RAVVJKCSZXAIQP-UHFFFAOYSA-N
methyl 5-bromovalerate,5-bromovaleric acid methyl ester,pentanoic acid, 5-bromo-, methyl ester,methyl .omega.-bromopentanoate,5-bromopentanoic acid methyl ester,5-bromo-pentanoic acid methyl ester,methyl omega-bromopentanoate,methyl5-bromovalerate,valeric acid, 5-bromo-, methyl ester,methyl-5-bromovalerate
79557
methyl 5-bromopentanoate
COC(=O)CCCCBr
Applications
It is employed as an intermediate in the production of Apixaban .

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Refractive Index 1.463
Density 1.363
Quantity 100g
Flash Point 99°C (210°F)
Boiling Point 79°C to 80°C (4mmHg)
Beilstein 1749649
Solubility Information Slightly soluble in water.
Formula Weight 195.06
Percent Purity 97%
Chemical Name or Material Methyl 5-bromovalerate

EINECSNumber : 226-709-7

TSCA : Yes

Recommended Storage : Ambient temperatures