Alfa Aesar™ Methyl 5-chloro-5-oxovalerate, 97%

Manufacturer: Alfa Aesar™ B2514118

Quantity 50g

 View more versions of this product

Catalog No. AAB2514118

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1501-26-4
    Molecular Formula C6H9ClO3
    Linear Formula ClCO(CH2)3CO2CH3
    Formula Weight 164.59
    MDL Number MFCD00000756
    UN Number UN3265
    Synonym Methyl 4-chloroformylbutyrate; Methyl glutaryl chloride
    Chemical Name or Material Methyl 5-chloro-5-oxovalerate

    Specifications

    Formula Weight 164.59
    Quantity 50g
    Recommended Storage 2° to 8°C
    Molecular Formula C6H9ClO3
    Linear Formula ClCO(CH2)3CO2CH3
    MDL Number MFCD00000756
    CAS 1501-26-4
    UN Number UN3265
    Synonym Methyl 4-chloroformylbutyrate; Methyl glutaryl chloride
    Health Hazard 1 Danger
    Boiling Point 108° to 110°C (15mmHg)
    Health Hazard 2 GHS H Statement
    H314-H318-H227
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Combustible liquid.
    Health Hazard 3 GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
    Chemical Name or Material Methyl 5-chloro-5-oxovalerate
    Assay 97%

    Description

    Applications
    Methyl 5-chloro-5-oxovalerate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

    Solubility
    Reacts with water.

    Notes
    Moisture sensitive. Store away from moisture and oxidizing agents under dry inert gas. Protect from humidity and water. Incompatible with oxidizing agents, bases and moisture.

    GHS H Statement
    H314-H318-H227
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Combustible liquid.

    2° to 8°C

    GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit