Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Methyl 5-chloropyrazine-2-carboxylate, 98%

Click to view available options
Quantity:
1g
5g
33332-25-1
C6H5ClN2O2
172.568
MFCD01632102
CVVMLRFXZPKILB-UHFFFAOYSA-N
methyl 5-chloro-2-pyrazinecarboxylate,methyl-5-chloropyrazine-2-carboxylate,2-chloro-5-carbomethoxy-pyrazine,5-chloro-pyrazine-2-carboxylic acid methyl ester,methyl-5-chloro-2-pyrazinecarboxylate,5-chloro-2-methoxycarbonyl pyrazine,2-pyrazinecarboxylic acid, 5-chloro-, methyl ester,pyrazinecarboxylic acid, 5-chloro-, methyl ester,5-chloro-2-pyrazinecarboxylic acid methyl ester,methyl-5-chlorpyrazin-2-carboxylat
406081
methyl 5-chloropyrazine-2-carboxylate
COC(=O)C1=CN=C(C=N1)Cl
33332-25-1
C6H5ClN2O2
172.568
MFCD01632102
CVVMLRFXZPKILB-UHFFFAOYSA-N
methyl 5-chloro-2-pyrazinecarboxylate,methyl-5-chloropyrazine-2-carboxylate,2-chloro-5-carbomethoxy-pyrazine,5-chloro-pyrazine-2-carboxylic acid methyl ester,methyl-5-chloro-2-pyrazinecarboxylate,5-chloro-2-methoxycarbonyl pyrazine,2-pyrazinecarboxylic acid, 5-chloro-, methyl ester,pyrazinecarboxylic acid, 5-chloro-, methyl ester,5-chloro-2-pyrazinecarboxylic acid methyl ester,methyl-5-chlorpyrazin-2-carboxylat
406081
methyl 5-chloropyrazine-2-carboxylate
COC(=O)C1=CN=C(C=N1)Cl
Quantity 5g
Beilstein 880046
Formula Weight 172.57
Physical Form Solid
Percent Purity 98%
Chemical Name or Material Methyl 5-chloropyrazine-2-carboxylate