Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 5-(chlorosulfonyl)-1-methyl-1H-pyrrole-2-carboxylate, 95%, Maybridge

Click to view available options
Quantity:
1g
Packaging:
Amber Glass Bottle
306936-53-8
C7H8ClNO4S
237.654
MFCD02180789
AKPDQZQBPBSNPS-UHFFFAOYSA-N
methyl 5-chlorosulfonyl-1-methyl-1h-pyrrole-2-carboxylate,methyl 5-chlorosulfonyl-1-methylpyrrole-2-carboxylate,methyl 5-chlorosulphonyl-1-methyl-1h-pyrrole-2-carboxylate,2-chlorosulphonyl-5-methoxycarbonyl-1-methyl-1h-pyrrole,5-ethoxycarbonyl-1-methyl-1h-pyrrole-2-sulphonyl chloride,5-chlorosulfonyl-1-methyl-2-pyrrolecarboxylic acid methyl ester,5-chlorosulfonyl-1-methyl-1h-pyrrole-2-carboxylic acid methyl ester,methyl 5-chlorosulphonyl-1-methylpyrrole-2-carboxylate,methyl-5-chlorosulfonyl-1-methyl-1h-pyrrole-2-carboxylate,1h-pyrrole-2-carboxylic acid, 5-chlorosulfonyl-1-methyl-, methyl ester 9ci
2736899
methyl 5-chlorosulfonyl-1-methylpyrrole-2-carboxylate
CN1C(=CC=C1S(=O)(=O)Cl)C(=O)OC
306936-53-8
C7H8ClNO4S
237.654
MFCD02180789
AKPDQZQBPBSNPS-UHFFFAOYSA-N
methyl 5-chlorosulfonyl-1-methyl-1h-pyrrole-2-carboxylate,methyl 5-chlorosulfonyl-1-methylpyrrole-2-carboxylate,methyl 5-chlorosulphonyl-1-methyl-1h-pyrrole-2-carboxylate,2-chlorosulphonyl-5-methoxycarbonyl-1-methyl-1h-pyrrole,5-ethoxycarbonyl-1-methyl-1h-pyrrole-2-sulphonyl chloride,5-chlorosulfonyl-1-methyl-2-pyrrolecarboxylic acid methyl ester,5-chlorosulfonyl-1-methyl-1h-pyrrole-2-carboxylic acid methyl ester,methyl 5-chlorosulphonyl-1-methylpyrrole-2-carboxylate,methyl-5-chlorosulfonyl-1-methyl-1h-pyrrole-2-carboxylate,1h-pyrrole-2-carboxylic acid, 5-chlorosulfonyl-1-methyl-, methyl ester 9ci
2736899
methyl 5-chlorosulfonyl-1-methylpyrrole-2-carboxylate
CN1C(=CC=C1S(=O)(=O)Cl)C(=O)OC
Quantity 1g
Packaging Amber Glass Bottle
CAS Min % 95.0
Assay Percent Range 95%
Formula Weight 237.66
Percent Purity 95%
Chemical Name or Material Methyl 5-(chlorosulfonyl)-1-methyl-1H-pyrrole-2-carboxylate

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger