Alfa Aesar™ Methyl 5-chlorovalerate, 96%

Manufacturer: Alfa Aesar™ B2540814

Quantity 25g

 View more versions of this product

Catalog No. AAB2540814

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 14273-86-0
    Molecular Formula C6ClH11O2
    Linear Formula Cl(CH2)4CO2CH3
    Formula Weight 150.61
    MDL Number MFCD00065274
    Synonym Methyl 5-chloropentanoate; 5-Chlorovaleric acid methyl ester
    Chemical Name or Material Methyl 5-chlorovalerate

    Specifications

    Formula Weight 150.61
    Molecular Formula C6ClH11O2
    Linear Formula Cl(CH2)4CO2CH3
    MDL Number MFCD00065274
    Synonym Methyl 5-chloropentanoate; 5-Chlorovaleric acid methyl ester
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335-H227
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.
    Health Hazard 3 GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Flash Point 90°C (194°F)
    Boiling Point 106° to 107°C (38mmHg)
    Quantity 25g
    CAS 14273-86-0
    Chemical Name or Material Methyl 5-chlorovalerate
    Assay 96%

    GHS H Statement
    H315-H319-H335-H227
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.

    GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit