Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 5-hydroxypyrazine-2-carboxylate, 97%, Thermo Scientific™

Click to view available options
Quantity:
1g
13924-95-3
C6H6N2O3
154.13
MFCD08273469,MFCD22683218
VNFXNPFSTYYFCZ-UHFFFAOYSA-N
methyl 5-hydroxypyrazine-2-carboxylate, 5-hydroxypyrazine-2-carboxylic acid methyl ester, 5-hydroxy-pyrazine-2-carboxylic acid methyl ester, methyl 5-hydropyrazine-2-carboxylate, methyl 5-hydroxy-2-pyrazinecarboxylate, methyl 5-oxo-4,5-dihydro-2-pyrazinecarboxylate, pubchem9107, 5-hydroxy-pyrazine-2-carboxylicacidmethylester, cbi-bb zero/008486, 2-hydroxy-5-carbomethoxy-pyrazine
8027173
methyl 6-oxo-1H-pyrazine-3-carboxylate
COC(=O)C1=CNC(=O)C=N1
13924-95-3
C6H6N2O3
154.13
MFCD08273469,MFCD22683218
VNFXNPFSTYYFCZ-UHFFFAOYSA-N
methyl 5-hydroxypyrazine-2-carboxylate, 5-hydroxypyrazine-2-carboxylic acid methyl ester, 5-hydroxy-pyrazine-2-carboxylic acid methyl ester, methyl 5-hydropyrazine-2-carboxylate, methyl 5-hydroxy-2-pyrazinecarboxylate, methyl 5-oxo-4,5-dihydro-2-pyrazinecarboxylate, pubchem9107, 5-hydroxy-pyrazine-2-carboxylicacidmethylester, cbi-bb zero/008486, 2-hydroxy-5-carbomethoxy-pyrazine
8027173
methyl 6-oxo-1H-pyrazine-3-carboxylate
COC(=O)C1=CNC(=O)C=N1
Melting Point 178.0°C to 181.0°C
Assay Percent Range 96% min. (HPLC)
Infrared Spectrum Authentic
Packaging Glass Bottle
Quantity 1g
Formula Weight 154.13
Percent Purity 97%
Chemical Name or Material Methyl 5-hydroxypyrazine-2-carboxylate

GHS H Statement Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.

GHS Signal Word: Warning

RUO – Research Use Only