Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Methyl 5-iodosalicylate, 99%

Click to view available options
Quantity:
10g
50g
250g
4068-75-1
C8H7IO3
278.045
MFCD00016462
NRSWJTRJHPRZMH-UHFFFAOYSA-N
methyl 5-iodosalicylate,5-iodosalicylic acid methyl ester,2-hydroxy-5-iodobenzoic acid methyl ester,benzoic acid, 2-hydroxy-5-iodo-, methyl ester,methyl 2-hydroxy-5-iodobenzenecarboxylate,salicylic acid, 5-iodo-, methyl ester,methyl5-iodosalicylate,methyl-5-iodosalicylate,acmc-209jg0,methyl2-hydroxy-5-iodobenzoate
248910
methyl 2-hydroxy-5-iodobenzoate
COC(=O)C1=C(C=CC(=C1)I)O
4068-75-1
C8H7IO3
278.045
MFCD00016462
NRSWJTRJHPRZMH-UHFFFAOYSA-N
methyl 5-iodosalicylate,5-iodosalicylic acid methyl ester,2-hydroxy-5-iodobenzoic acid methyl ester,benzoic acid, 2-hydroxy-5-iodo-, methyl ester,methyl 2-hydroxy-5-iodobenzenecarboxylate,salicylic acid, 5-iodo-, methyl ester,methyl5-iodosalicylate,methyl-5-iodosalicylate,acmc-209jg0,methyl2-hydroxy-5-iodobenzoate
248910
methyl 2-hydroxy-5-iodobenzoate
COC(=O)C1=C(C=CC(=C1)I)O
Melting Point 75°C to 78°C
Quantity 50g
Formula Weight 278.05
Percent Purity 99%
Chemical Name or Material Methyl 5-iodosalicylate

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning