Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 5-iodosalicylate 99%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
Packaging:
Glass Bottle
4068-75-1
C8H7IO3
278.045
MFCD00016462
NRSWJTRJHPRZMH-UHFFFAOYSA-N
methyl 5-iodosalicylate,5-iodosalicylic acid methyl ester,2-hydroxy-5-iodobenzoic acid methyl ester,benzoic acid, 2-hydroxy-5-iodo-, methyl ester,methyl 2-hydroxy-5-iodobenzenecarboxylate,salicylic acid, 5-iodo-, methyl ester,methyl5-iodosalicylate,methyl-5-iodosalicylate,acmc-209jg0,methyl2-hydroxy-5-iodobenzoate
248910
methyl 2-hydroxy-5-iodobenzoate
COC(=O)C1=C(C=CC(=C1)I)O
4068-75-1
C8H7IO3
278.045
MFCD00016462
NRSWJTRJHPRZMH-UHFFFAOYSA-N
methyl 5-iodosalicylate,5-iodosalicylic acid methyl ester,2-hydroxy-5-iodobenzoic acid methyl ester,benzoic acid, 2-hydroxy-5-iodo-, methyl ester,methyl 2-hydroxy-5-iodobenzenecarboxylate,salicylic acid, 5-iodo-, methyl ester,methyl5-iodosalicylate,methyl-5-iodosalicylate,acmc-209jg0,methyl2-hydroxy-5-iodobenzoate
248910
methyl 2-hydroxy-5-iodobenzoate
COC(=O)C1=C(C=CC(=C1)I)O
Packaging Glass Bottle
Quantity 10g
Melting Point 73°C to 78°C
Infrared Spectrum Authentic
Color White
Assay Percent Range 99%
Linear Formula IC6H3(OH)COOCH3
Formula Weight 278.04
Physical Form Crystalline Powder
Percent Purity ≥98.5%
Chemical Name or Material Methyl 5-iodosalicylate
Show More Show Less

GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.