Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 5-methylsalicylate 97%, ACROS Organics™

Click to view available options
Quantity:
25g
22717-57-3
C9H10O3
166.176
MFCD00016485
JQYUQKRFSSSGJM-UHFFFAOYSA-N
methyl 5-methylsalicylate,benzoic acid, 2-hydroxy-5-methyl-, methyl ester,2-hydroxy-5-methylbenzoic acid methyl ester,methyl-5-methylsalicylate,2,5-cresotic acid, methyl ester,methyl 5-methyl salicylate,p-cresotic acid methyl ester,5-methylsalicylic acid methyl ester,methyl 2-hydroxy-5-methyl-benzoate,6-hydroxy-m-toluic acid methyl ester
89802
methyl 2-hydroxy-5-methylbenzoate
CC1=CC(=C(C=C1)O)C(=O)OC
22717-57-3
C9H10O3
166.176
MFCD00016485
JQYUQKRFSSSGJM-UHFFFAOYSA-N
methyl 5-methylsalicylate,benzoic acid, 2-hydroxy-5-methyl-, methyl ester,2-hydroxy-5-methylbenzoic acid methyl ester,methyl-5-methylsalicylate,2,5-cresotic acid, methyl ester,methyl 5-methyl salicylate,p-cresotic acid methyl ester,5-methylsalicylic acid methyl ester,methyl 2-hydroxy-5-methyl-benzoate,6-hydroxy-m-toluic acid methyl ester
89802
methyl 2-hydroxy-5-methylbenzoate
CC1=CC(=C(C=C1)O)C(=O)OC
Quantity 25g
Color Yellow
Refractive Index 1.5320 to 1.5340 (20°C, 589nm)
Infrared Spectrum Authentic
Assay Percent Range 97%
Linear Formula CH3C6H3(OH)COOCH3
Formula Weight 166.18
Physical Form Liquid
Percent Purity ≥96.0%
Chemical Name or Material Methyl 5-methylsalicylate, 97%

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 245-173-5

TSCA : TSCA