Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 5-nitro-2-furoate, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
1874-23-3
C6H5NO5
171.108
MFCD00051956
UTLKCGPAJUYGOM-UHFFFAOYSA-N
methyl 5-nitro-2-furoate,methyl nitrofuroate,methyl 5-nitrofuroate,5-nitro-2-furoic acid methyl ester,methyl 5-nitropyromucate,methyl 5-nitro-2-furancarboxylate,unii-71qm4e6j90,ccris 2152,5-nitro-2-furancarboxylic acid methyl ester,2-furancarboxylic acid, 5-nitro-, methyl ester
94882
methyl 5-nitrofuran-2-carboxylate
COC(=O)C1=CC=C(O1)[N+](=O)[O-]
1874-23-3
C6H5NO5
171.108
MFCD00051956
UTLKCGPAJUYGOM-UHFFFAOYSA-N
methyl 5-nitro-2-furoate,methyl nitrofuroate,methyl 5-nitrofuroate,5-nitro-2-furoic acid methyl ester,methyl 5-nitropyromucate,methyl 5-nitro-2-furancarboxylate,unii-71qm4e6j90,ccris 2152,5-nitro-2-furancarboxylic acid methyl ester,2-furancarboxylic acid, 5-nitro-, methyl ester
94882
methyl 5-nitrofuran-2-carboxylate
COC(=O)C1=CC=C(O1)[N+](=O)[O-]
Melting Point 78°C to 83°C
Infrared Spectrum Authentic
Color Yellow
Quantity 25g
Packaging Glass bottle
Assay Percent Range 97%
Formula Weight 171.11
Physical Form Crystalline Powder
Percent Purity ≥96.0%
Chemical Name or Material Methyl 5-nitro-2-furoate, 97%

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RTECSNumber : LV2013000