Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl-5-norbornene-2,3-dicarboxylic anhydride, 95%, mixture of isomers, ACROS Organics™

Click to view available options
Quantity:
250g
1kg
5kg
Packaging:
Glass bottle
Plastic drum
25134-21-8
C10H10O3
178.18
MFCD00167036
KNRCVAANTQNTPT-UHFFFAOYNA-N
Methyl nadic anhydride,Methylbicyclo[2.2.1]hept-5-ene-2, 3-dicarboxylic anhydride
25134-21-8
C10H10O3
178.18
MFCD00167036
KNRCVAANTQNTPT-UHFFFAOYNA-N
Methyl nadic anhydride,Methylbicyclo[2.2.1]hept-5-ene-2, 3-dicarboxylic anhydride
Viscosity 220-300 mPa.s (25°C)
Infrared Spectrum Authentic
Boiling Point 275.0°C
Packaging Glass bottle
Flash Point 140°C
Quantity 250g
Refractive Index 1.5060 to 1.5080
Color Yellow
Specific Gravity 1.23
Density 1.2300g/mL
Assay Percent Range 94% min. (GC)
Beilstein 17, 461
Solubility Information Solubility in water: <1g/L
Formula Weight 178.18
Physical Form Oily Liquid
Percent Purity 95%
Chemical Name or Material Methyl-5-norbornene-2, 3-dicarboxylic anhydride, Mixture of isomers
Show More Show Less

GHS H Statement
Causes serious eye damage.
Toxic if inhaled.
Harmful if swallowed.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
Causes skin irritation

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 246-644-8