Alfa Aesar™ Methyl-5-norbornene-2,3-dicarboxylic anhydride, mixture of isomers, tech.

Manufacturer: Alfa Aesar™ A1098222

Quantity 100g

 View more versions of this product

Catalog No. AAA1098222

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 25134-21-8
    Molecular Formula C10H10O3
    Formula Weight 178.19
    MDL Number MFCD00167036
    Synonym Methyl nadic anhydride
    Chemical Name or Material Methyl-5-norbornene-2,3-dicarboxylic anhydride, Mixture of isomers
    Grade Technical Grade

    Specifications

    Formula Weight 178.19
    Health Hazard 3 GHS P Statement
    P284a-P261-P301+P310-P305+P351+P338-P405-P501a
    Wear respiratory protection.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H301-H334-H332-H315-H319-H335
    Toxic if swallowed.
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Synonym Methyl nadic anhydride
    Quantity 100g
    TSCA TSCA
    Molecular Formula C10H10O3
    MDL Number MFCD00167036
    CAS 25134-21-8
    Health Hazard 1 Danger
    Boiling Point 137° to 140°C (10mmHg)
    Flash Point 135°C (275°F)
    Chemical Name or Material Methyl-5-norbornene-2,3-dicarboxylic anhydride, Mixture of isomers
    Grade Technical Grade

    Description

    Applications
    Methyl-5-norbornene-2,3-dicarboxylic anhydride is used as a curing agent in epoxy resins and in the preparation of vertically aligned and penetrated carbon nanotube/polymer film. It serves as an intermediate in polyesters, alkyd resins and plasticizers. It is also used in electrical laminating and filament-winding. Further, it is used in the preparation of methyl nadimides endcapped resins based on tris(3-aminophenyl)phosphine oxide. It is utilized in the flame retarding treatment.

    Solubility
    Miscible with acetone, benzene, naphtha and xylene.

    Notes
    Moisture sensitive. Incompatible with strong oxidizing agents, strong acids and strong bases.

    GHS H Statement
    H301-H334-H332-H315-H319-H335
    Toxic if swallowed.
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P284a-P261-P301+P310-P305+P351+P338-P405-P501a
    Wear respiratory protection.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit