Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Methyl 6-bromo-2-naphthoate, 98%

Click to view available options
Quantity:
1g
5g
25g
33626-98-1
C12H9BrO2
265.106
MFCD00100408
JEUBRLPXJZOGPX-UHFFFAOYSA-N
methyl 6-bromo-2-naphthoate,methyl6-bromo-2-naphthoate,6-bromo-2-naphthalenecarboxylic acid methyl ester,6-bromo-2-naphthoic acid methyl ester,2-naphthalenecarboxylic acid, 6-bromo-, methyl ester,6-bromonaphthalene-2-carboxylic acid methyl ester,6-bromo-naphthalene-2-carboxylic acid methyl ester,pubchem9467,acmc-209i1z,6-bromo-2-methylnaphthoate
854134
methyl 6-bromonaphthalene-2-carboxylate
COC(=O)C1=CC2=C(C=C1)C=C(C=C2)Br
33626-98-1
C12H9BrO2
265.106
MFCD00100408
JEUBRLPXJZOGPX-UHFFFAOYSA-N
methyl 6-bromo-2-naphthoate,methyl6-bromo-2-naphthoate,6-bromo-2-naphthalenecarboxylic acid methyl ester,6-bromo-2-naphthoic acid methyl ester,2-naphthalenecarboxylic acid, 6-bromo-, methyl ester,6-bromonaphthalene-2-carboxylic acid methyl ester,6-bromo-naphthalene-2-carboxylic acid methyl ester,pubchem9467,acmc-209i1z,6-bromo-2-methylnaphthoate
854134
methyl 6-bromonaphthalene-2-carboxylate
COC(=O)C1=CC2=C(C=C1)C=C(C=C2)Br
Quantity 1g
Melting Point 121°C to 123°C
Beilstein 2578943
Formula Weight 265.11
Percent Purity 98%
Chemical Name or Material Methyl 6-bromo-2-naphthoate