Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Methyl 6-chloropyridine-2-carboxylate, 95%

Click to view available options
Quantity:
5g
25g
100g
6636-55-1
C7H6ClNO2
171.58
MFCD00155391
TWUXBVMXSBEKHA-UHFFFAOYSA-N
methyl 6-chloropicolinate,methyl 6-chloro-2-pyridinecarboxylate,2-pyridinecarboxylic acid, 6-chloro-, methyl ester,2-chloropyridine-6-carboxylate methyl,6-chloro-2-picolinic acid methyl ester,6-chloropicolinic acid methyl ester,methyl 2-chloro-6-pyridinecarboxylate,methyl 2-chloropyridine-6-carboxylate,6-chloro-pyridine-2-carboxylic acid methyl ester,6-chloro-2-pyridinecarboxylic acid methyl ester
242723
methyl 6-chloropyridine-2-carboxylate
COC(=O)C1=NC(=CC=C1)Cl
6636-55-1
C7H6ClNO2
171.58
MFCD00155391
TWUXBVMXSBEKHA-UHFFFAOYSA-N
methyl 6-chloropicolinate,methyl 6-chloro-2-pyridinecarboxylate,2-pyridinecarboxylic acid, 6-chloro-, methyl ester,2-chloropyridine-6-carboxylate methyl,6-chloro-2-picolinic acid methyl ester,6-chloropicolinic acid methyl ester,methyl 2-chloro-6-pyridinecarboxylate,methyl 2-chloropyridine-6-carboxylate,6-chloro-pyridine-2-carboxylic acid methyl ester,6-chloro-2-pyridinecarboxylic acid methyl ester
242723
methyl 6-chloropyridine-2-carboxylate
COC(=O)C1=NC(=CC=C1)Cl
Applications
Methyl 6-chloropyridine-2-carboxylate was coupled with 2-(trimethylstannyl)pyridine to afford methyl 2,2′-bipyridine-6-carboxylate.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, heat.
Color Yellow
Melting Point 92°C to 98°C (literature)
Quantity 100g
Beilstein 128580
Solubility Information Slightly soluble in water.
Formula Weight 171.58
Physical Form Solid
Percent Purity 95%
Chemical Name or Material Methyl 6-chloropyridine-2-carboxylate

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C