Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl acetate, 99+%, Extra Dry, AcroSeal™, Acros Organics

Click to view available options
Quantity:
100mL
1L
2.5L
79-20-9
C3H6O2
74.08
MFCD00008711
KXKVLQRXCPHEJC-UHFFFAOYSA-N
tereton,devoton,acetic acid, methyl ester,methyl ethanoate,acetic acid methyl ester,methylacetat,acetate de methyle,methylacetaat,methyl acetic ester,octan metylu
6584
CHEBI:77700
methyl acetate
CC(=O)OC
79-20-9
C3H6O2
74.08
MFCD00008711
KXKVLQRXCPHEJC-UHFFFAOYSA-N
tereton,devoton,acetic acid, methyl ester,methyl ethanoate,acetic acid methyl ester,methylacetat,acetate de methyle,methylacetaat,methyl acetic ester,octan metylu
6584
CHEBI:77700
methyl acetate
CC(=O)OC
Viscosity 0.38 mPa.s (20°C)
Packaging AcroSeal glass Bottle
Density 0.9300g/mL
Infrared Spectrum Authentic
Boiling Point 57.4°C
Color Colorless to Yellow
Melting Point -98.0°C
Specific Gravity 0.93
Water 0.005% Max.
Refractive Index 1.3600 to 1.3620
Quantity 100mL
Residue after Evaporation 0.0005 % max.
Flash Point −10°C
Assay Percent Range 99% min. (GC)
Linear Formula CH3CO2CH3
Beilstein 02, 224
Merck Index 15, 6082
Solubility Information Solubility in water: 250g/L (20°C). Other solubilities: miscible with alcohol and ether
Formula Weight 74.08
Physical Form Solution
Percent Purity 99+%
Grade Extra Dry
Chemical Name or Material Methyl acetate
Show More Show Less

GHS H Statement:
May cause drowsiness or dizziness.
Causes serious eye irritation.
Highly flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 201-185-2