Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl acetate, 99%, extra pure, ACROS Organics™

Click to view available options
Quantity:
1L
2.5L
10L
Packaging:
Glass Bottle
Plastic drum
79-20-9
C3H6O2
74.08
MFCD00008711
KXKVLQRXCPHEJC-UHFFFAOYSA-N
tereton,devoton,acetic acid, methyl ester,methyl ethanoate,acetic acid methyl ester,methylacetat,acetate de methyle,methylacetaat,methyl acetic ester,octan metylu
6584
CHEBI:77700
methyl acetate
CC(=O)OC
79-20-9
C3H6O2
74.08
MFCD00008711
KXKVLQRXCPHEJC-UHFFFAOYSA-N
tereton,devoton,acetic acid, methyl ester,methyl ethanoate,acetic acid methyl ester,methylacetat,acetate de methyle,methylacetaat,methyl acetic ester,octan metylu
6584
CHEBI:77700
methyl acetate
CC(=O)OC
Viscosity 0.38 mPa.s (20°C)
Quantity 10L
Packaging Plastic drum
Specific Gravity 0.93
Boiling Point 57.4°C
Infrared Spectrum Authentic
Residue after Evaporation 0.001% max.
Melting Point -98.0°C
Color Undesignated
Water 0.05% max. (K.F.)
Density 0.9300g/mL
Flash Point −10°C
Refractive Index 1.3600 to 1.3620
Assay Percent Range 98.5% min. (GC)
Linear Formula CH3CO2CH3
Beilstein 02, 224
Merck Index 15, 6082
Solubility Information Solubility in water: 250g/L (20°C). Other solubilities: miscible with alcohol and ether
Formula Weight 74.08
Physical Form Liquid
Percent Purity 99%
Grade Extra Pure
Chemical Name or Material Methyl acetate
Show More Show Less

GHS H Statement:
May cause drowsiness or dizziness.
Causes serious eye irritation.
Highly flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 201-185-2