Alfa Aesar™ Methyl acetopyruvate, 98%

Manufacturer: Alfa Aesar™ A1119014

Quantity 25g

 View more versions of this product

Catalog No. AAA1119014

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 20577-61-1
    Molecular Formula C8H8O4
    Formula Weight 144.13
    MDL Number MFCD00043638
    Synonym Acetopyruvic acid methyl ester; Methyl 2,4-dioxovalerate
    Chemical Name or Material Methyl acetopyruvate

    Specifications

    Formula Weight 144.13
    Quantity 25g
    Molecular Formula C8H8O4
    MDL Number MFCD00043638
    Melting Point 60° to 63°C
    Synonym Acetopyruvic acid methyl ester; Methyl 2,4-dioxovalerate
    CAS 20577-61-1
    Chemical Name or Material Methyl acetopyruvate
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit