Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl acrylate, ≥99.5% (GC), MilliporeSigma™ Supelco™

$57.50 - $157.90

Specifications

Percent Purity ≥99.5% (GC)
Density 0.956 g/mL (at 25°C)
Grade Analytical Standard
Linear Formula CH2=CHCOOCH3
Physical Form Neat
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
11-101-9988
View Documents
MilliporeSigma Supelco
767781MLF
1 mL
Each for $57.50
Add to cart
 
11-101-8698
View Documents
MilliporeSigma Supelco
767785MLF
5 mL
Each for $157.90
Add to cart
 
Description

Description

Environmental and Petroleum
Specifications

Specifications

≥99.5% (GC)
Analytical Standard
Neat
0.956 g/mL (at 25°C)
CH2=CHCOOCH3
Safety and Handling

Safety and Handling

ShelfLife : Limited shelf life, expiry date on the label

RTECSNumber : AT2800000

SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit