Alfa Aesar™ Methyl-alpha-D-mannopyranoside, 99%

Manufacturer: Alfa Aesar™ A1153322

Quantity 100g

 View more versions of this product

Catalog No. AAA1153322

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 617-04-9
    Molecular Formula C7H14O6
    Formula Weight 194.18
    MDL Number MFCD00063262
    Beilstein 81566
    Chemical Name or Material Methyl-alpha-D-mannopyranoside

    Specifications

    Formula Weight 194.18
    Beilstein 81566
    Molecular Formula C7H14O6
    MDL Number MFCD00063262
    CAS 617-04-9
    Melting Point 193° to 196°C
    Quantity 100g
    Optical Rotation +79° (c=10 in water)
    Chemical Name or Material Methyl-alpha-D-mannopyranoside
    Assay 99%

    Description

    Applications
    Methyl a-D-mannopyranoside Is a competitor inhibitor of the binding of mannose by Escherichia coli. It is also used as a pharmaceutical raw material, health care product and intermediate. It is also been used as Animal Pharmaceuticals.

    Solubility
    It is easily soluble in methanol, water. Insoluble in ether.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agent. Stable under recommended storage conditions

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit