Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Anthranilate 99%, ACROS Organics™

Click to view available options
Quantity:
50g
Packaging:
Glass bottle
134-20-3
C8H9NO2
151.165
MFCD00007710
VAMXMNNIEUEQDV-UHFFFAOYSA-N
methyl anthranilate,2-aminobenzoic acid methyl ester,anthranilic acid methyl ester,methyl o-aminobenzoate,nevoli oil,methylanthranilate,o-carbomethoxyaniline,2-carbomethoxyaniline,2-methoxycarbonyl aniline,benzoic acid, 2-amino-, methyl ester
8635
CHEBI:73244
methyl 2-aminobenzoate
COC(=O)C1=CC=CC=C1N
134-20-3
C8H9NO2
151.165
MFCD00007710
VAMXMNNIEUEQDV-UHFFFAOYSA-N
methyl anthranilate,2-aminobenzoic acid methyl ester,anthranilic acid methyl ester,methyl o-aminobenzoate,nevoli oil,methylanthranilate,o-carbomethoxyaniline,2-carbomethoxyaniline,2-methoxycarbonyl aniline,benzoic acid, 2-amino-, methyl ester
8635
CHEBI:73244
methyl 2-aminobenzoate
COC(=O)C1=CC=CC=C1N
Infrared Spectrum Authentic
Packaging Glass bottle
Color Brown-Yellow
Quantity 50g
Refractive Index 1.5810 to 1.5840 (20°C, 589nm)
Density 1.1680g/mL
Assay Percent Range 99%
Linear Formula 2-(H2N)C6H4CO2CH3
Beilstein 14, 317
Merck Index 15, 6094
Formula Weight 151.16
Physical Form Liquid
Percent Purity ≥98.5%
Chemical Name or Material Methyl anthranilate, 99%
Show More Show Less

GHS P Statement Causes serious eye irritation. Causes skin irritation.

GHS P Statement IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact le

Warning

EINECSNumber : 205-132-4

RTECSNumber : CB3325000

TSCA : TSCA