Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl arachidate, 99%, Acros Organics

Click to view available options
Quantity:
1g
5g
25g
1120-28-1
C21H42O2
326.56
QGBRLVONZXHAKJ-UHFFFAOYSA-N
methyl arachidate,eicosanoic acid, methyl ester,arachidic acid methyl ester,methyl eicosanoate,methyl arachisate,arachidic acid methylester,eicosanoic acid methyl ester,methylarachidate,methyl aracidate
14259
methyl icosanoate
CCCCCCCCCCCCCCCCCCCC(=O)OC
1120-28-1
C21H42O2
326.56
QGBRLVONZXHAKJ-UHFFFAOYSA-N
methyl arachidate,eicosanoic acid, methyl ester,arachidic acid methyl ester,methyl eicosanoate,methyl arachisate,arachidic acid methylester,eicosanoic acid methyl ester,methylarachidate,methyl aracidate
14259
methyl icosanoate
CCCCCCCCCCCCCCCCCCCC(=O)OC
Packaging Glass Bottle
Quantity 1g
Flash Point >110°C
Melting Point 45.0°C to 48.0°C
Boiling Point 215.0°C to 216.0°C (10.0mmHg)
Assay Percent Range 98.5 % min. (GC)
Linear Formula CH3(CH2)18COOCH3
Formula Weight 326.56
Percent Purity 99%
Chemical Name or Material Methyl arachidate

EINECSNumber : 214-304-8