Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl benzoate, 99%, ACROS Organics™

Click to view available options
Quantity:
500mL
2.5L
10L
Packaging:
Glass bottle
Plastic drum
93-58-3
C8H8O2
136.15
QPJVMBTYPHYUOC-UHFFFAOYSA-N
methylbenzoate,benzoic acid, methyl ester,clorius,benzoic acid methyl ester,niobe oil,oil of niobe,methyl benzenecarboxylate,essence of niobe,oniobe oil,oxidate le
7150
CHEBI:72775
methyl benzoate
COC(=O)C1=CC=CC=C1
93-58-3
C8H8O2
136.15
QPJVMBTYPHYUOC-UHFFFAOYSA-N
methylbenzoate,benzoic acid, methyl ester,clorius,benzoic acid methyl ester,niobe oil,oil of niobe,methyl benzenecarboxylate,essence of niobe,oniobe oil,oxidate le
7150
CHEBI:72775
methyl benzoate
COC(=O)C1=CC=CC=C1
Viscosity 1.94 mPa.s (23°C)
Melting Point -12.0°C
Color Colorless to Yellow
Boiling Point 198.0°C to 199.0°C
Infrared Spectrum Authentic
Acidity 1mg KOH/g max.
Density 1.0800g/mL
Refractive Index 1.5160 to 1.5180
Flash Point 77°C
Specific Gravity 1.08
Quantity 500mL
Packaging Glass bottle
Assay Percent Range 98.5% min. (GC)
Linear Formula C6H5CO2CH3
Beilstein 09,109
Merck Index 15,6098
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with alcohol,methanol and ether
Formula Weight 136.15
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material Methyl benzoate
Show More Show Less

GHS H Statement
Harmful if swallowed.

GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

GHS Signal Word: Warning

EINECSNumber : 202-259-7