Alfa Aesar™ (Methyl benzoate)tricarbonylchromium, 98%

Catalyst for hydrogenation of unsaturated fats

Manufacturer: Alfa Aesar™ 4436806

Quantity 5g

 View more versions of this product

Catalog No. AA4436806

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12125-87-0
    Molecular Formula C11H8CrO5
    Formula Weight 272.18
    MDL Number MFCD00009957
    UN Number UN3466
    Synonym (Methyl benzoate)chromium tricarbonyl
    Chemical Name or Material (Methyl benzoate)tricarbonylchromium

    Specifications

    Formula Weight 272.18
    Health Hazard 3 GHS P Statement
    P264-P270-P301+P310-P321-P405-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Specific treatment (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00009957
    Quantity 5g
    Sensitivity Air & Moisture Sensitive
    Health Hazard 2 GHS H Statement
    H300
    Fatal if swallowed.
    Molecular Formula C11H8CrO5
    Synonym (Methyl benzoate)chromium tricarbonyl
    Color Orange
    Melting Point 95° to 96°C
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: METAL CARBONYLS, SOLID, N.O.S.
    Physical Form Crystalline
    CAS 12125-87-0
    UN Number UN3466
    Health Hazard 1 Danger
    Chemical Name or Material (Methyl benzoate)tricarbonylchromium
    Assay 98%

    GHS H Statement
    H300
    Fatal if swallowed.

    GHS P Statement
    P264-P270-P301+P310-P321-P405-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Specific treatment (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit