Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl-beta-cyclodextrin, average Mw 1310, Acros Organics

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
128446-36-6
C54H94O35
1303.311
MFCD00074980
YZOUYRAONFXZSI-SBHWVFSVSA-N
beta-cyclodextrin methyl ethers,beta-cyd,methyl-?cyclodextrin,beta-w7 m1.8,dimethyl-,a-cyclodextrin,.beta.-cyclodextrin,methylethers,methyl-b-cyclodextrin cell culture*tested
51051622
COC1C(C2C(OC1OC3C(OC(C(C3OC)OC)OC4C(OC(C(C4OC)OC)OC5C(OC(C(C5OC)OC)OC6C(OC(C(C6OC)OC)OC7C(OC(C(C7OC)OC)OC8C(OC(O2)C(C8O)OC)CO)CO)CO)CO)CO)CO)CO)O
128446-36-6
C54H94O35
1303.311
MFCD00074980
YZOUYRAONFXZSI-SBHWVFSVSA-N
beta-cyclodextrin methyl ethers,beta-cyd,methyl-?cyclodextrin,beta-w7 m1.8,dimethyl-,a-cyclodextrin,.beta.-cyclodextrin,methylethers,methyl-b-cyclodextrin cell culture*tested
51051622
COC1C(C2C(OC1OC3C(OC(C(C3OC)OC)OC4C(OC(C(C4OC)OC)OC5C(OC(C(C5OC)OC)OC6C(OC(C(C6OC)OC)OC7C(OC(C(C7OC)OC)OC8C(OC(O2)C(C8O)OC)CO)CO)CO)CO)CO)CO)CO)O
Chelation/Complexation reagent
Loss on Drying 5.0% max.
Infrared Spectrum Authentic
Specific Rotation +161° to +167° (20°C, 589nm) (C=5, water)
Color White
Flash Point 187°C
Quantity 25g
Melting Point 165.0°C to 172.0°C
Density 187g/mL
Packaging Glass bottle
Solubility Information Solubility in water: 800g/L (25°C). Other solubilities: soluble in methanol,ethanol,acetone,pyridine,,dimethylsulfoxide and dimethylformamide
Physical Form Powder
Chemical Name or Material Methyl-β-cyclodextrin, Average Mn approx. 1310
Show More Show Less