Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Methyl-beta-cyclodextrin, average Mw 1310, Acros Organics
Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
Description
Chelation/Complexation reagent
Specifications
Loss on Drying | 5.0% max. |
Flash Point | 187°C |
Quantity | 100g |
Packaging | Glass bottle |
Melting Point | 165.0°C to 172.0°C |
Infrared Spectrum | Authentic |
Specific Rotation | +161° to +167° (20°C, 589nm) (C=5, water) |
Color | White |
Density | 187g/mL |
Solubility Information | Solubility in water: 800g/L (25°C). Other solubilities: soluble in methanol,ethanol,acetone,pyridine,,dimethylsulfoxide and dimethylformamide |
Show More |