Alfa Aesar™ Methyl bromoacetate, 98+%

Manufacturer: Alfa Aesar™ A1060518

Quantity 50g

 View more versions of this product

Catalog No. AAA1060518

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 96-32-2
    Molecular Formula C3H5BrO2
    Linear Formula BrCH2CO2CH3
    Formula Weight 152.98
    MDL Number MFCD00000189
    UN Number UN2643
    Beilstein 506256
    Synonym Bromoacetic acid methyl ester
    Chemical Name or Material Methyl bromoacetate

    Specifications

    Formula Weight 152.98
    Health Hazard 3 GHS P Statement
    P210-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula C3H5BrO2
    Quantity 50g
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: METHYL BROMOACETATE
    Linear Formula BrCH2CO2CH3
    MDL Number MFCD00000189
    Melting Point −50°C
    Health Hazard 1 Danger
    RTECS Number AF6300000
    TSCA TSCA
    CAS 96-32-2
    Beilstein 506256
    Flash Point 62°C (143°F)
    Health Hazard 2 GHS H Statement
    H301-H311-H331-H314-H318-H227
    Toxic if swallowed.
    Toxic in contact with skin.
    Toxic if inhaled.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Combustible liquid.
    UN Number UN2643
    Synonym Bromoacetic acid methyl ester
    Boiling Point 144° to 145°C
    Odor Pungent
    Chemical Name or Material Methyl bromoacetate
    Assay ≥98%

    Description

    Applications
    Methyl bromoacetate is used in the synthesis of coumarins and cis-cyclopropanes. It reacts with the conjugate base of (methylmethoxycarbene)pentacarbonylchromium(0) to prepare alkylated carbene complexes. Further, it is used to make vitamins and pharmaceuticals.

    Solubility
    Miscible with methanol, ether and acetone. Slightly miscible with water

    Notes
    Incompatible with acids, bases, oxidizing agents and reducing agents.

    GHS H Statement
    H301-H311-H331-H314-H318-H227
    Toxic if swallowed.
    Toxic in contact with skin.
    Toxic if inhaled.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Combustible liquid.

    GHS P Statement
    P210-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit