Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl α-bromophenylacetate, 97+%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
3042-81-7
C9H9BrO2
229.07
MFCD00013535
NHFBYYMNJUMVOT-UHFFFAOYSA-N
methyl alpha-bromophenylacetate,methyl bromophenylacetate,methyl bromo phenyl acetate,methyl .alpha.-bromophenylacetate,methyl2-bromo-2-phenylacetate,methyl .alpha.-bromobenzeneacetate,benzeneacetic acid, .alpha.-bromo-, methyl ester,+/--methyl alpha-bromophenylacetate,alpha-bromophenylacetic acid methyl ester,methyl .alpha.-bromo-.alpha.-phenylacetate
137806
methyl 2-bromo-2-phenylacetate
COC(=O)C(C1=CC=CC=C1)Br
3042-81-7
C9H9BrO2
229.07
MFCD00013535
NHFBYYMNJUMVOT-UHFFFAOYSA-N
methyl alpha-bromophenylacetate,methyl bromophenylacetate,methyl bromo phenyl acetate,methyl .alpha.-bromophenylacetate,methyl2-bromo-2-phenylacetate,methyl .alpha.-bromobenzeneacetate,benzeneacetic acid, .alpha.-bromo-, methyl ester,+/--methyl alpha-bromophenylacetate,alpha-bromophenylacetic acid methyl ester,methyl .alpha.-bromo-.alpha.-phenylacetate
137806
methyl 2-bromo-2-phenylacetate
COC(=O)C(C1=CC=CC=C1)Br
Quantity 1g
Flash Point 113°C
Boiling Point 113.0°C (3.0 mmHg)
Density 1.4550g/mL
Packaging Glass bottle
Specific Gravity 1.455
Assay Percent Range 97% min. (GC)
Linear Formula C6H5CH(Br)CO2CH3
Solubility Information Solubility in water: .. Other solubilities: soluble in most organic solvents
Formula Weight 229.07
Percent Purity 97+%
Chemical Name or Material Methyl α-bromophenylacetate
Show More Show Less

GHS Signal Word: Danger