Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl caprate, 98+%, Acros Organics

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
110-42-9
C11H22O2
186.29
MFCD00009580
YRHYCMZPEVDGFQ-UHFFFAOYSA-N
methyl caprate,decanoic acid, methyl ester,methyl caprinate,capric acid methyl ester,decanoic acid methyl ester,methyl-n-caprate,uniphat a30,metholene 2095,methyl n-decanoate,methyl n-caprate
8050
methyl decanoate
CCCCCCCCCC(=O)OC
110-42-9
C11H22O2
186.29
MFCD00009580
YRHYCMZPEVDGFQ-UHFFFAOYSA-N
methyl caprate,decanoic acid, methyl ester,methyl caprinate,capric acid methyl ester,decanoic acid methyl ester,methyl-n-caprate,uniphat a30,metholene 2095,methyl n-decanoate,methyl n-caprate
8050
methyl decanoate
CCCCCCCCCC(=O)OC
Boiling Point 226°C
Melting Point -18°C
Flash Point 90°C
Specific Gravity 0.871
Quantity 500g
Refractive Index 1.4240 to 1.4270 (20°C, 589nm)
Density 0.8710g/mL
Infrared Spectrum Authentic
Packaging Glass bottle
Color Colorless
Assay Percent Range 98% min. (GC)
Linear Formula CH3(CH2)8CO2CH3
Solubility Information Solubility in water: practically insoluble
Formula Weight 186.29
Physical Form Liquid
Percent Purity 98+%
Chemical Name or Material Methyl caprate
Show More Show Less

GHS H Statement
Toxic to aquatic life with long lasting effects.

GHS P Statement
Avoid release to the environment.

Warning

EINECSNumber : 203-766-6