Alfa Aesar™ Methyl cellulose, viscosity 1600 cPs

Manufacturer: Alfa Aesar™ 4314636

Quantity 500g

 View more versions of this product

Catalog No. AA4314636

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 9004-67-5
    MDL Number MFCD00081763
    Merck Index 14,6040
    Chemical Name or Material Methyl cellulose, Viscosity: 1600cPs

    Specifications

    TSCA TSCA
    Physical Form Powder
    MDL Number MFCD00081763
    Odor Odorless
    Quantity 500g
    CAS 9004-67-5
    Merck Index 14,6040
    Chemical Name or Material Methyl cellulose, Viscosity: 1600cPs

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit