Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Methyl cyclopentene-3-carboxylate, 97%

Click to view available options
Quantity:
1g
5g
58101-60-3
C7H10O2
126.155
MFCD04038661
CEOILRYKIJRPBZ-UHFFFAOYSA-N
methyl 3-cyclopentenecarboxylate,methyl cyclopent-3-enecarboxylate,methyl-3-cyclopentene-1-carboxylate,methyl3-cyclopentenecarboxylate,3-cyclopentene-1-carboxylic acid methyl ester,3-cyclopentene-1-carboxylic acid, methyl ester,acmc-209m2j,methyl cyclopent-3-ene carboxylate,4-methoxycarbonyl cyclopent-1-ene,methyl 3-cyclopentene-1-carboxylate
1514118
methyl cyclopent-3-ene-1-carboxylate
COC(=O)C1CC=CC1
58101-60-3
C7H10O2
126.155
MFCD04038661
CEOILRYKIJRPBZ-UHFFFAOYSA-N
methyl 3-cyclopentenecarboxylate,methyl cyclopent-3-enecarboxylate,methyl-3-cyclopentene-1-carboxylate,methyl3-cyclopentenecarboxylate,3-cyclopentene-1-carboxylic acid methyl ester,3-cyclopentene-1-carboxylic acid, methyl ester,acmc-209m2j,methyl cyclopent-3-ene carboxylate,4-methoxycarbonyl cyclopent-1-ene,methyl 3-cyclopentene-1-carboxylate
1514118
methyl cyclopent-3-ene-1-carboxylate
COC(=O)C1CC=CC1
Color Brown to Colorless
Refractive Index 1.4500
Density 1.03g/mL
Quantity 5g
UN Number UN1993
Formula Weight 126.15
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material Methyl cyclopentene-3-carboxylate

GHS H Statement
H318-H226-H335-H315
Causes serious eye damage.
Flammable liquid and vapor.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packaging Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.