Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl-α-D-galactopyranoside, 98+%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
3396-99-4
C7H14O6
194.183
MFCD00064085
HOVAGTYPODGVJG-PZRMXXKTSA-N
methyl alpha-d-galactopyranoside,alpha-methyl-d-galactoside,alpha-methylgalactoside,me-alpha-gal,methyl alpha-d-galactoside,2r,3r,4s,5r,6s-2-hydroxymethyl-6-methoxytetrahydro-2h-pyran-3,4,5-triol,methyl,a-d-galactopyranoside,methyl-galactopyranoside,methyl-alpha-d-galactopyranoside,alpha-methyl d-galactoside
76935
CHEBI:55507
(2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol
COC1C(C(C(C(O1)CO)O)O)O
3396-99-4
C7H14O6
194.183
MFCD00064085
HOVAGTYPODGVJG-PZRMXXKTSA-N
methyl alpha-d-galactopyranoside,alpha-methyl-d-galactoside,alpha-methylgalactoside,me-alpha-gal,methyl alpha-d-galactoside,2r,3r,4s,5r,6s-2-hydroxymethyl-6-methoxytetrahydro-2h-pyran-3,4,5-triol,methyl,a-d-galactopyranoside,methyl-galactopyranoside,methyl-alpha-d-galactopyranoside,alpha-methyl d-galactoside
76935
CHEBI:55507
(2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol
COC1C(C(C(C(O1)CO)O)O)O
Specific Rotation Condition + 179.00 (20.00°C c=1,water)
Quantity 10g
Assay Percent Range 98+%
Formula Weight 194.18
Chemical Name or Material Methyl-α-D-galactopyranoside