Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Methyl decanoate, 99%

Click to view available options
Quantity:
50g
100g
250g
110-42-9
C11H22O2
186.295
MFCD00009580
YRHYCMZPEVDGFQ-UHFFFAOYSA-N
methyl caprate,decanoic acid, methyl ester,methyl caprinate,capric acid methyl ester,decanoic acid methyl ester,methyl-n-caprate,uniphat a30,metholene 2095,methyl n-decanoate,methyl n-caprate
8050
methyl decanoate
CCCCCCCCCC(=O)OC
110-42-9
C11H22O2
186.295
MFCD00009580
YRHYCMZPEVDGFQ-UHFFFAOYSA-N
methyl caprate,decanoic acid, methyl ester,methyl caprinate,capric acid methyl ester,decanoic acid methyl ester,methyl-n-caprate,uniphat a30,metholene 2095,methyl n-decanoate,methyl n-caprate
8050
methyl decanoate
CCCCCCCCCC(=O)OC
Applications
It is employed as an intermediate for caproic acid, detergents, emulsifiers, wetting agents, stabilizers, resins, lubricants, plasticizers and flavoring. As a chemical intermediate for alkanolamides, fatty alcohols & acids.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Flash Point 94°C (201°F)
Boiling Point 223°C to 225°C
Quantity 250g
Refractive Index 1.425
Density 0.877
Melting Point -18°C
Beilstein 1759170
Solubility Information Soluble in alcohol. Insoluble in water.
Formula Weight 186.3
Percent Purity 99%
Chemical Name or Material Methyl decanoate
Show More Show Less

P210-P233-P240-P241-P242-P243-P280-P374-P501c

H227

EINECSNumber : 203-766-6

RTECSNumber : HD9830000

TSCA : Yes

Recommended Storage : Ambient temperatures