Alfa Aesar™ Methyl diphenylphosphinite, 98+%

Manufacturer: Alfa Aesar™ A1155522

Quantity 100g

 View more versions of this product

Catalog No. AAA1155522

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 4020-99-9
    Molecular Formula C13H13OP
    Formula Weight 216.22
    MDL Number MFCD00048025
    Beilstein 1640480
    Synonym Diphenylphosphinous acid methyl ester
    Chemical Name or Material Methyl diphenylphosphinite

    Specifications

    Formula Weight 216.22
    Health Hazard 3 GHS P Statement
    P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Take off contaminated clothing and wash before reuse.
    Specific treatment (see label).
    If skin irritation occurs: Get medical advice/attention.
    If eye irritation persists: Get medical advice/attention.
    MDL Number MFCD00048025
    CAS 4020-99-9
    Beilstein 1640480
    Synonym Diphenylphosphinous acid methyl ester
    Health Hazard 1 Warning
    Boiling Point 148° to 150°C (6mmHg)
    Quantity 100g
    Recommended Storage −30°C to −10°C
    Molecular Formula C13H13OP
    Flash Point >110°C (230°F)
    Health Hazard 2 GHS H Statement
    H315-H319
    Causes skin irritation.
    Causes serious eye irritation.
    Chemical Name or Material Methyl diphenylphosphinite
    Assay ≥98%

    GHS H Statement
    H315-H319
    Causes skin irritation.
    Causes serious eye irritation.

    −30°C to −10°C

    GHS P Statement
    P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Take off contaminated clothing and wash before reuse.
    Specific treatment (see label).
    If skin irritation occurs: Get medical advice/attention.
    If eye irritation persists: Get medical advice/attention.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit